VWR® Durac™, Baumé hydrometer
Supplier: VWR Collection
34670-140
VWRU34670-140EA 54.8 SGD
VWRU34670-140
VWR® Durac™, Baumé hydrometer
Hydrometers
Baumé hydrometer designed for determining the specific gravity and Baumé in liquids lighter then water.
  • Dual scale
  • Individually serialised
  • Lead free metal ballast
Specially used in chemistry, pharmacology, food and beverage, and in the starch industry.
Certifications: NIST and DAkkS. Calibration conforms to A2LA accreditions.
Delivery information: Supplied with a multi-language statement of accuracy.