VWR®Signature™, Low temperature bod refrigerated incubators
Supplier: VWR Collection
35960-056 35960-057
VWRU35960-056EA 11050 SGD
VWRU35960-056 VWRU35960-057
VWR®Signature™, Low temperature bod refrigerated incubators
Incubators BOD Incubators
These refrigerated incubators with microprocessor control are widely used for applications such as BOD determinations, plant and insect studies, fermentation studies and bacterial culturing.
  • CFC-free refrigerant
  • Digital display of temperature
  • Easy keypad set and calibration
  • Safety high/low-limit thermostat
  • Over/under counter
  • Gentle forced air circulation
  • Interior electrical outlet
Units have a hermetically-sealed compressor, a circuit breaker to protect from electrical overload and an easy-to-clean, fully insulated chamber. Gentle, continuous forced-More Product Information