VWR®Signature™, Incubators, reach-in
Supplier: VWR Collection
35962-096
VWRU35962-096EA 28860 SGD
VWRU35962-096
VWR®Signature™, Incubators, reach-in
Incubators Standard Incubators
These incubators provide gentle air flow and minimises the space. And the chamber reinforced to support roller apparatus and shakers.
  • Microprocessor temperature control
  • Over temperature safety protection
  • Temperature uniformity to 0,8 °C
  • Prewired access panel for chart recorder
The air jacket design and heated outer door allows for uniform heat throughout the incubators. Both models incorporate a microprocessor controller to achieve precise temperature uniformity. An independent, secondary temperature controller offers the added security of overMore Product Information