VWR® Dust'it™, Blower
Supplier: VWR Collection
36934-142
VWRU36934-142EA 44.8 SGD
VWRU36934-142
VWR® Dust'it™, Blower
Sprays
These blowers are ideal for delicate, difficult-to-clean, and inaccessible equipments. The nonconductive plastic sleeve tip prevents scratches for cleaning optical lenses, slides, circuit boards, and cuvets.
  • Blows away dust, lint, and dirt
  • Environmentally safe
  • Powerful air-jet
  • One-way inlet valve at the top keeps air from being drawn in
Order Now
SPECIFICATIONS
Environmentally Preferable