VWR® Traceable®, Digital thermometer, platinum freezer
Supplier: VWR Collection
36934-160
VWRU36934-160EA 315 SGD
VWRU36934-160
VWR® Traceable®, Digital thermometer, platinum freezer
Thermometers Digital Thermometers
RTD platinum freezer thermometer which is used to monitor the temperature in freezers, water baths, heating blocks, incubators and refrigerators.
  • Probe with thin microcable supplies high-precision results
  • Ultra-slim micro-10' cable allows freezer doors to seal tightly
  • Adjustable probe holder, magnets, Velcro® allow unit/probe to be used anywhere in lab/field
Probe dimension (Ø×L): 4,1×76,2 mm
Certifications: Compliance with ISO/IEC 17025; accredited by A2LA indicating traceability to standards provided by NIST.
Delivery information: SuppliedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable