VWR® Microtube Storage Boxes and Racks
Supplier: VWR Collection
40000-332 40000-342 40000-320 40000-322 40000-340 40000-324 40000-326 40000-328 40000-330
VWRU40000-332EA 129 SGD
VWRU40000-332 VWRU40000-342 VWRU40000-320 VWRU40000-322 VWRU40000-340 VWRU40000-324 VWRU40000-326 VWRU40000-328 VWRU40000-330
VWR® Microtube Storage Boxes and Racks
Boxes Cryoboxes
These durable, polypropylene boxes maintain their integrity under harsh freezer conditions and will not absorb water or degrade like cardboard storage boxes.
Each box holds one hundred 1,5 or 2,0 ml tubes. A three-point hinge and positive catch provide quick access to samples and keep them secure when box is closed. Boxes are available in seven colours (with clear lids). An alphanumeric grid on the lid aids in sample identification. Autoclavable to 121 °C.
Ordering information: Assorted boxes includes one box in each of the following colours: blue, green, orange, pink, and yellow.