VWR® CERTICLEAN® Cleanroom Gloves, Nitrile, Class 10
Supplier: VWR Collection
40101-352 40101-354 40101-356
VWRU40101-352EA 0 SGD
VWRU40101-352 VWRU40101-354 VWRU40101-356
VWR® CERTICLEAN® Cleanroom Gloves, Nitrile, Class 10
Gloves
Ambidextrous gloves with beaded cuffs made from silicone-free nitrile. The specialised manufacturing processes result in consistently low particle counts and low levels of ionic extractables and non-volatile residue (NVR) transfer.
  • Non sterile, powder-free, textured fingertips
  • In-use static dissipative properties are superior to natural rubber latex
  • Tapered cuff provides more coverage over the garment sleeve
Suitable for use in Class 10, ISO 4, M2.5 and lower cleanrooms.
Packaging: Packaged in cleanroom-compatible polyethylene, 100 gloves/doubleMore Product Information