VWR® Spectrosil®, Fluorometer cuvettes
Supplier: VWR Collection
414004-064 414004-057
VWRU414004-064EA 1110 SGD
VWRU414004-064 VWRU414004-057
VWR® Spectrosil®, Fluorometer cuvettes
Cuvettes Spectrophotometer Cuvettes
Standard rectangular cells manufactured from Spectrosil® quartz for accurate readings over the entire UV range to near infrared range.
  • Far UV quartz
  • Wavelength range of 170 to 2700 nm
  • Cells exhibit a transmission tolerance within 1,5% at 200 nm
Due to the fully heat-fused construction (no intermediate materials or adhesives) and careful annealing to remove any strain, there is no optical or physical distortion of the four optically polished windows.
Lids and stoppers are made of polytetrafluoroethylene (PTFE) except cell 414004-057 which features aMore Product Information