VWR®, Spectrophotometer Cuvettes
Supplier: VWR Collection
414004-066
VWRU414004-066EA 229 SGD
VWRU414004-066
VWR®, Spectrophotometer Cuvettes
Cuvettes Spectrophotometer Cuvettes
Accurate and dependable results for spectrophotometric analyses in the visible and near infrared regions.
  • Matched sets within the accepted pathlength standard of ±0,01 mm
  • Cells exhibit a transmission tolerance within 0,5% at 365 nm
  • Lids and stoppers are made of PTFE
Due to the fully heat-fused construction (no intermediate materials or adhesives) and careful annealing to remove any strain, there is no optical or physical distortion of the two optical windows.