VWR®, Bottles, Narrow Neck, Round, LDPE, with Screw Caps
Supplier: VWR Collection
414004-184
VWRU414004-184EA 50.1 SGD
VWRU414004-184
VWR®, Bottles, Narrow Neck, Round, LDPE, with Screw Caps
Bottles Plastic Bottles
LDPE, translucent.
  • Leakproof, robust and versatile
  • Excellent chemical resistance
  • Built-in loop on the shoulder for identification tag attachment
Excellent for sample collection, storing and transporting liquids.