VWR®, Bottles, Wide Neck, with Screw Cap
Supplier: VWR Collection
414004-187
VWRU414004-187EA 36.7 SGD
VWRU414004-187
VWR®, Bottles, Wide Neck, with Screw Cap
Bottles Plastic Bottles
PP, translucent.
These bottles are specifically designed for applications that require autoclaving of large volumes. They are not leakproof and are not recommended for shipping liquids.