VWR®, Wash Bottles
Supplier: VWR Collection
414004-227
VWRU414004-227EA 45.2 SGD
VWRU414004-227
VWR®, Wash Bottles
Bottles Wash Bottles
These LDPE wash bottle features a PP screw cap and stem that are molded into a one-piece unit for leakproof operation.
Dispensing tube produces a fine stream, but can be cut for a coarser flow.