VWR® Cryo Cooler® , Freezing containers
Supplier: VWR Collection
414004-284
VWRU414004-284EA 157 SGD
VWRU414004-284
VWR® Cryo Cooler® , Freezing containers
Boxes Cooling/Freezing Boxes and Bags
Polycarbonate cooler, with floating inserts.
  • Non-mechanical
  • Withstands repeated use
  • Holds upto eighteen 1,2 to 2 ml cryogenic vials
These coolers provides the –1 °C per minute cooling rate required to successfully preserve cells, when filled with isopropyl alcohol and placed in mechanical freezer.