VWR® Advanced Lab Coats
Supplier: VWR Collection
414004-375
VWRU414004-375EA 149 SGD
VWRU414004-375
VWR® Advanced Lab Coats
Disposable Garments
Lab coats are manufactured from a specially formulated breathable microporous fabric that is exceptionally clean and durable.
  • Significant fluid and particulate barrier
  • Durable lightweight fabric
  • Excellent breathability and water vapor transmission rate
  • Soft tactile feel
These low-linting garments provide a fluid barrier and dry-particulate protection, as well as resistance to common laboratory chemicals. Appropriate for industrial, pharmaceutical, food processing, construction, and certain environmental cleanup applications.
Garments featureMore Product Information