VWR®, Protective gowns, SPP
Supplier: VWR Collection
414004-468
VWRU414004-468EA 84.7 SGD
VWRU414004-468
VWR®, Protective gowns, SPP
Disposable Garments
Economical gowns are manufactured from a strong, lightweight, breathable and non-linting spunbonded polypropylene (SPP) fabric.
  • Provide first line barrier
  • Elastic cuffs and ties at the neck and waist ensure secure closure with
  • Serged seams
  • No pockets
Gowns protect against dirt, grime and certain dry particulates in nonhazardous environments.
Certifications: Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.
Order Now
SPECIFICATIONS
Environmentally Preferable
Fabric SPP
Pockets Without pockets
Gender
Seam type Serged
Garment application Medical,Clinical