VWR® Maximum, Gowns
Supplier: VWR Collection
414004-470
VWRU414004-470EA 206 SGD
VWRU414004-470
VWR® Maximum, Gowns
Disposable Garments
These gowns are the cleanest and most durable within the entire VWR® Protection line of apparel. Gowns provide barrier protection against fluids, biologicals, particulates, and light chemical splash.
  • Full protection from fluids and light chemical splash
  • Manufactured from fluid-impervious material
  • Lowest level of particle shedding
  • Clean, low-linting fabric
  • Elastic cuffs and ties at the neck and waist ensure secure closure
  • Serged seams
  • Bacterial filtration efficiency: 99,9%
Appropriate for industrial, pharmaceutical, food processing, and certainMore Product Information