VWR®, Sleeve Protectors
Supplier: VWR Collection
414004-510
VWRU414004-510EA 485 SGD
VWRU414004-510
VWR®, Sleeve Protectors
Disposable Garments
These sterile sleeve covers are made from a specially formulated breathable microporous fabric that provides significant fluid and barrier protection. The sleeves feature elastic openings and sonically welded seams.
  • Gamma irradiated to a Sterility Assurance Level (SAL) of 10⁻⁶
  • Lowest level of particle shedding: Level I Helmke Drum Classification
  • Durable fabric with excellent breathability and water vapor transmission rate
  • Significant fluid and particulate barrier: 99,9% bacterial filtration efficiency
  • Soft, cloth-like fabric
Manufactured and packagedMore Product Information