VWR®, Cleanroom Wipes, Non Woven
Supplier: VWR Collection
414004-515
VWRU414004-515EA 733 SGD
VWRU414004-515
VWR®, Cleanroom Wipes, Non Woven
Wipes Cleanroom Wipes Dry Cleanroom Wipes
This wipe is made from a hydroentangled blend (45% polyester and 55% cellulose) that combines the strength and cleanliness of synthetic polyester with the high absorbency of cellulose.
  • Soft, economical
  • Very precise, knife-cut edges
  • No chemical binders
Designed for cleaning surfaces and controlling spills in ISO Class 6 (Class 1000/M4.5) and less critical environments.
Certifications: Complies with RoHS 2002/95/EC.
Packaging: Vacuum-packed in solvent-safe, cleanroom-compatible PE with a label on each inner bag.