VWR®, Drying tubes
Supplier: VWR Collection
46600-050
VWRU46600-050EA 44.5 SGD
VWRU46600-050
VWR®, Drying tubes
Tubes Drying Tubes
PE, translucent. Suitable for drying gases, these tapered tubes have two push-on ends for easily filling with drying agents. The tapered connections accept tubes with diameters of 8, 9, and 10 mm.