VWR® Spilltray™, Workstation and drying rack
Supplier: VWR Collection
46610-022
VWRU46610-022EA 83.5 SGD
VWRU46610-022
VWR® Spilltray™, Workstation and drying rack
Trays/Dishes/Pots Spillage Trays/Pans
PE, containment tray.
  • Easy to clean
  • Chemical-resistant and maintenance-free
  • Easel design permits drying of odd shaped items
  • Plastic grid cushions and protects against breakage and scratches
  • Handles safely chemicals and liquids while pipeting, measuring or mixing
  • Entire tray is dishwasher safe
Dimension: 419,1×330×16 mm
Weight: 0,45 kg