VWR® Traceable®, Infrared tachometer
Supplier: VWR Collection
46610-048
VWRU46610-048EA 430 SGD
VWRU46610-048
VWR® Traceable®, Infrared tachometer
Tachometers
This touchless laser tachometer is designed to check speeds of centrifuges, pumps, rollers, shafts, gears, motors or stirrers.
  • Touchless operation for accurate readings in two seconds
  • Optical non contact measurements, impact resistant housing
  • Memory feature to recall highest, lowest and last reading
Certifications: ISO/IEC 17025:2005 (1750.01) accredited by A2LA indicating traceability to standards provided by NIST.
Delivery information: Supplied with a nine-volt battery, 762 mm of reflective tape and Traceable® certificate.