VWR® Traceable®, Radio-controlled wall clock
Supplier: VWR Collection
47732-424
VWRU47732-424EA 281 SGD
VWRU47732-424
VWR® Traceable®, Radio-controlled wall clock
Timers Wall Clocks
Controlling radio signal from NIST (National Institute of Standards and Technology) in Colorado automatically adjusts clock to reflect precise time, daylight savings, leap year, and leap seconds (earth rotation wobble corrections).
  • 57,2 mm (²¹/₄") high digits for visibility from 6,1 m (20') distance
  • Clock displays time in a.m./p.m. or 24-hour time, month, date, day of week, and ambient temperature from –0 to 50 °C (32 to 122 °F)
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA toMore Product Information