VWR®, Ring Weights for Flasks, C-Shaped
Supplier: VWR Collection
47735-598 47735-600 47735-596 47735-594
VWRU47735-598EA 98.3 SGD
VWRU47735-598 VWRU47735-600 VWRU47735-596 VWRU47735-594
VWR®, Ring Weights for Flasks, C-Shaped
Weights Flask Weights
Lead core, with Vikem® vinyl coating. These lead rings fit neatly around the neck of filtering flasks with side-arm tubing connections without disconnecting the tubing.
  • Stabilise vessels in water baths or on the benchtop
  • Open ring is flexible enough to be pulled apart to accommodate a wider vessel