VWR® ColorFrost, Microscope Slides
Supplier: VWR Collection
48312-502
VWRU48312-502EA 46 SGD
VWRU48312-502
VWR® ColorFrost, Microscope Slides
Slides Microscope Slides
Slides frosted with a brilliantly coloured, opaque coating, which takes and retains most marking methods.
  • Easy to write on and to read: Key shows which side is up
  • No sticking or scratching, coating impervious to chemicals and reagents
  • Slides with raised printed writing surface that eliminates paper labels and re-labelling
Order Now
SPECIFICATIONS
Environmentally Preferable