VWR® SuperSlips™, Micro Cover Glasses, Rectangular
Supplier: VWR Collection
48404-455
VWRU48404-455EA 19.2 SGD
VWRU48404-455
VWR® SuperSlips™, Micro Cover Glasses, Rectangular
Cover Glasses
These cover glasses are corrosion-resistant and uniformly thin and flat.
  • Suitable for humid environments
  • Resistant to corrosion
  • Processing techniques result in superior cleanliness
SuperSlip coverslips combine specialised production methods with moisture-resistant packaging to prevent sticking.
Order Now
SPECIFICATIONS
Environmentally Preferable