VWR®, Marker pen
Supplier: VWR Collection
52877-310 52877-398 52877-355 52877-435
VWRU52877-310EA 49.5 SGD
VWRU52877-310 VWRU52877-398 VWRU52877-355 VWRU52877-435
VWR®, Marker pen
Pens Lab Markers
Special-purpose markers write through moisture as easily as they write on dry surfaces, suitable for industrial, biomedical, and educational laboratories. Markers provide permanent marking on plastic disposables. When used on glass or metal reusables, or on porcelain surfaces, ink will not smear when subjected to water and can be washed off these surfaces using various solvents, allowing repeated marking.
  • Provide permanent marking on plastic disposables
  • Dries quickly for immediate handling of labware
  • Durable tip and generous ink supply provide a long product life