VWR®, Culture tube, disposable, with screw cap finish
Supplier: VWR Collection
53283-686 53283-804 53283-685 53283-800 53283-802
VWRU53283-686EA 520 SGD
VWRU53283-686 VWRU53283-804 VWRU53283-685 VWRU53283-800 VWRU53283-802
VWR®, Culture tube, disposable, with screw cap finish
Tubes Culture Tubes
Borosilicate glass 3.3, disposable.
Tubes have a marking spot and reference mark for orientation during use, and a threaded neck to accept screw cap.
Ordering information: Caps must be ordered separately.
Order Now
SPECIFICATIONS
Environmentally Preferable