VWR®, Pipette Filler

Supplier: VWR Collection
53502-205
VWRU53502-205EA 45.4 SGD
VWRU53502-205
VWR®, Pipette Filler
Pipettes Pipette Bulbs
Pipette bulb made of synthetic rubber.

  • For use with pipettes from 2 to 50 ml
  • Made of synthetic rubber with glass ball valves
  • No metal parts to corrode
  • Supplied with an adapter for capacities greater than 20 ml
Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR