VWR® Pipette Pump™ , Pipetting devices
Supplier: VWR Collection
53502-233 53502-244
VWRU53502-233EA 60.6 SGD
VWRU53502-233 VWRU53502-244
VWR® Pipette Pump™ , Pipetting devices
Pipettes Pipette Controllers
These pipetting devices accept both disposable and standard pipettes in the flexible tapered chuck, per ASTM Standard E714-80. They provide safe, accurate and trouble-free pipetting with simple, one-handed operation.
  • Pipettes fit smoothly into its flexible tapered chuck
  • Thumbwheel on side is rotated for precision aspirating or dispensing
  • Plunger may be pressed for quick emptying
  • Resistant to acids and alkalis
  • Disassemble all parts for cleaning and before autoclaving at 121 °C (250 °F) for 20 minutes; chuck should not be autoclaved
  • Colour-coded for quick
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable