VWR®CryoPro®, Cryogenic storage tanks
Supplier: VWR Collection
55709-218 55709-224
VWRU55709-218EA 0 SGD
VWRU55709-218 VWRU55709-224
VWR®CryoPro®, Cryogenic storage tanks
Dewars and Cryogenic Vessels
For small to medium capacity applications, the CC series offers traditional canister-style cryogenic storage in efficient narrow neck designs.
CryoRetriever claw-style tool is specifically designed for retrieving boxes, vials, and frames from cryopreservation tanks. Exclusive sealed center shaft prevents hazardous spurting of LN₂ that can occur with a hollow shaft retrieval claw. Aluminium canes simplify and organise cryogenic vials for storage in LN₂ dewars with canisters. One cane holds six 1,2 ml to 2 ml vials.
Sturdy five-wheel rollerbases are available for safely moving full tanks.More Product Information