VWR®CryoPro®, Liquid nitrogen dewars, l-series
Supplier: VWR Collection
55709-240 55709-236 55709-234 82021-112
VWRU55709-240EA 3750 SGD
VWRU55709-240 VWRU55709-236 VWRU55709-234 VWRU82021-112
VWR®CryoPro®, Liquid nitrogen dewars, l-series
Dewars and Cryogenic Vessels
The L series liquid dewars designed for efficient and safe handling of liquid nitrogen. Stores and dispenses liquid nitrogen safely. With super insulated liquid dewars provide a minimal evaporation rates and are available for pouring, pressurised dispensing and dipping of liquid nitrogen.