VWR®, Bottle Carrier
Supplier: VWR Collection
56608-864
VWRU56608-864EA 248 SGD
VWRU56608-864
VWR®, Bottle Carrier
Carriers Bottle Carriers/Containers
These bottle carriers made of HDPE are suitable for transport and storage of reagent bottles. The cone-shaped opening allows a delivery tube to be inserted, so liquid does not have to be poured out.
  • Epoxy coating
  • Heavy-gauge steel wire bail handle
  • Side ribs and bottom suspend the filled bottle
  • Colour: White
Order Now
SPECIFICATIONS
Environmentally Preferable