VWR® SoftCIDE™, Antibacterial hand cleansers
Supplier: VWR Collection
56614-420
VWRU56614-420EA 89.6 SGD
VWRU56614-420
VWR® SoftCIDE™, Antibacterial hand cleansers
Skincare Soaps and Skin Cleansers
Extra-mild, fast-acting, antimicrobial soap with 1% PCMX. Specialised formulations and mild ingredients help to replenish the natural oils of the skin.
  • Effective against a broad spectrum micro-organism
  • pH balanced with the skin
  • Triclosan-free