VWR®, Spectrophotometer Cuvettes
Supplier: VWR Collection
58017-875
VWRU58017-875EA 301 SGD
VWRU58017-875
VWR®, Spectrophotometer Cuvettes
Cuvettes Spectrophotometer Cuvettes
Cuvette manufactured in clear materials to support different wavelength range and pathlength requirements.
  • Recessed windows
  • Suitable for accurate UV visible analyses
  • Four optical windows recessed to prevent scratching