VWR®Spinbar®, Magnetic stirring bars, disposable
Supplier: VWR Collection
58947-140
VWRU58947-140EA 219 SGD
VWRU58947-140
VWR®Spinbar®, Magnetic stirring bars, disposable
Stirrers Magnetic Stirring Bars
The multifaceted stirring bar surface adds turbulence relative to similar size smooth cylindrical stirring bars.
  • Ideal for single use applications such as mixing radioactive reagents
  • Disposable; eliminates potential for cross-contamination
  • Alnico magnet encapsulated in FDA grade Teflon® PTFE is durable and inert
For single-use application where efficiency and cross-contamination are a concern. Alnico V magnet.