VWR®Spinplus®, Magnetic stirring bars
Supplier: VWR Collection
58947-822 58947-828 58947-830 58947-820 58947-826
VWRU58947-822EA 9.2 SGD
VWRU58947-822 VWRU58947-828 VWRU58947-830 VWRU58947-820 VWRU58947-826
VWR®Spinplus®, Magnetic stirring bars
Stirrers Magnetic Stirring Bars
These magnetic stirring bars add speed and efficiency to mixing operations. The '+' shape creates a deep vortex and ensures stable and quiet operation.
  • PTFE resin-coated
Ordering information: Order each size separately or choose a five-piece assortment (one of each size).
Order Now
SPECIFICATIONS
Environmentally Preferable