VWR® Spinbar®, Magnetic stirring bars, micro
Supplier: VWR Collection
58948-069 58948-375 58948-397 58948-353
VWRU58948-069EA 27.1 SGD
VWRU58948-069 VWRU58948-375 VWRU58948-397 VWRU58948-353
VWR® Spinbar®, Magnetic stirring bars, micro
Stirrers Magnetic Stirring Bars
Mainly designed for stirring small volumes in vessels such as vials, tubes and gradient makers.
  • Available in a variety of colours and sizes
  • Used in environmental testing and life science applications