VWR®, Magnetic stirring bars, elliptical
Supplier: VWR Collection
58949-008 58949-016 58949-210 58949-010 58949-012
VWRU58949-008EA 39.3 SGD
VWRU58949-008 VWRU58949-016 VWRU58949-210 VWRU58949-010 VWRU58949-012
VWR®, Magnetic stirring bars, elliptical
Stirrers Magnetic Stirring Bars
PTFE covered elliptical magnetic stirring bars, except the three smallest sizes which have a cylindrical magnet in an egg-shaped PTFE coating. These elliptical stirring bars, designed especially for use in round-bottom vessels, spin unrestricted by the vessel contour. Autoclavable.
  • Very good chemical resistance
  • Provide their own pivot point without a separate ring or abrupt change in contour
  • Offer minimal contact when used in plastic containers
Order Now
SPECIFICATIONS
Environmentally Preferable