VWR® Spinvane®, Magnetic stirring bars
Supplier: VWR Collection
58949-270 58949-272
VWRU58949-270EA 8.8 SGD
VWRU58949-270 VWRU58949-272
VWR® Spinvane®, Magnetic stirring bars
Stirrers Magnetic Stirring Bars
PTFE covered, Alnico V magnetic core. Spinvane® Triangle shaped magnetic stirring bars are designed for stirring in test tubes, centrifuge tubes and micro vials.
  • The wings can also be trimmed to size if needed
  • Autoclave safe