VWR®, Stoppers, Neoprene, Green, without Holes
Supplier: VWR Collection
59589-336
VWRU59589-336EA 172 SGD
VWRU59589-336
VWR®, Stoppers, Neoprene, Green, without Holes
Stoppers Rubber Stoppers
These stoppers contain no free sulphur. It exhibits good resilency and chemical resistance.
  • Neoprene
  • Green