VWR® Talon® , Lab frames, small
Supplier: VWR Collection
60075-008
VWRU60075-008EA 1000 SGD
VWRU60075-008
VWR® Talon® , Lab frames, small
Retort Rods
These vertical mounting lab frames are ideal for glassware set up in small laboratories or where space is limited. Frames measure 610×610 mm (24×24") with a 457 mm (18") wide base for stability. Base may be permanently mounted to benchtop with screws (included).
Base dimension: 457 mm
Frame dimensions: 610×610 mm
Delivery information: Each lab frame includes eight 51 mm rods, two 457 mm rods, eight 610 mm rods, four rod end connectors, eight S connectors and four lab frame feet.
Order Now
SPECIFICATIONS
Environmentally Preferable