VWR® Talon® , Rod end connector
Supplier: VWR Collection
60079-010
VWRU60079-010EA 24.5 SGD
VWRU60079-010
VWR® Talon® , Rod end connector
Retort Rods
This nickel-plated zinc connector holds rods firmly at 90°.
  • Use this rod end connector when semi-permanent installations are required
  • Comes with two set screws and is precision bored for close fit with 13 mm (¹/₂") rods
Order Now
SPECIFICATIONS
Environmentally Preferable