VWR® Talon®, Lab frame feet
Supplier: VWR Collection
60079-257
VWRU60079-257EA 44.8 SGD
VWRU60079-257
VWR® Talon®, Lab frame feet
Retort Stands
Strong, durable lab frame foot made of nickel plated zinc. Designed for mounting lab frames permanently to bench tops, fume hoods, walls, and floors.
  • Accepts 13 mm (¹/₂") rods
  • Spilled fluids roll right off the smooth body contour, minimising contamination and allowing easy clean-ups
  • Features a set screw and includes three 19 mm (³/₄") mounting screws
  • Stainless steel lab frame foot includes stainless steel hardware
Order Now
SPECIFICATIONS
Environmentally Preferable