VWR® Talon®, Support rods, stainless steel
Supplier: VWR Collection
60079-535 60079-537 60079-534
VWRU60079-535EA 95.1 SGD
VWRU60079-535 VWRU60079-537 VWRU60079-534
VWR® Talon®, Support rods, stainless steel
Retort Rods
Stainless steel 303.
  • Heavy duty, maximum strength and durability
  • For use with VWR® Talon® lab-frames
Order Now
SPECIFICATIONS
Environmentally Preferable