VWR® Talon®, Lab frames, medium
Supplier: VWR Collection
60079-554
VWRU60079-554EA 3130 SGD
VWRU60079-554
VWR® Talon®, Lab frames, medium
Retort Stands
This lab frame with horizontal or vertical mounting is ideal for distillation and general set-ups.
  • With stainless steel rods
  • Can be assembled as 1220×610 mm horizontal or 610×1220 mm vertical frame
  • Base is 460 mm wide for stability
Delivery information: Each lab frame includes eight 51 mm rods, two 457 mm rods, seven 610 mm rods, four 1220 mm rods, four rod end connectors, thirty five S-connectors and four lab frame feet.