VWR®, Adhesive PCR Film Seal
Supplier: VWR Collection
60941-070
VWRU60941-070EA 195 SGD
VWRU60941-070
VWR®, Adhesive PCR Film Seal
Seals Microplate Seals
These heat resistant 50 µm thick films are designed for thermal cycling applications. Polypropylene films are not pierceable. For PCR applications where piercing with pipette tips or robotic probes is required for product recovery, use aluminium foil films. For Real-Time PCR applications where maximum optical clarity is required, use optically clear polyester films.
  • Recommended for temperatures from −40 to +120 °C
  • Certified free from DNase, RNase and nuclease
Each film L×D: 135,1×79,4 mm with sufficient sealing area for all PCR plates
Length withMore Product Information