VWR®, Polyester Films for ELISA and General Incubation
Supplier: VWR Collection
60941-120
VWRU60941-120EA 127 SGD
VWRU60941-120
VWR®, Polyester Films for ELISA and General Incubation
Seals Microplate Seals
These 50 μm thick polyester general-purpose films are designed for ELISA, EIA, and incubation. Films feature a single end tab with no perforations and are suppled non sterile.
Each film L×D: 133×83 mm