VWR®, Draining Racks
Supplier: VWR Collection
60996-012
VWRU60996-012EA 728 SGD
VWRU60996-012
VWR®, Draining Racks
Racks Draining Racks
PS.
  • Lightweight and versatile
  • With 76 mm draining ledge
  • Pegs snap out easily
This rack accommodates cylinders, flasks and test tubes.