VWR® Traceable® Humidity Sponge™, Dessicants
Supplier: VWR Collection
61161-319 61161-321
VWRU61161-319EA 178 SGD
VWRU61161-319 VWRU61161-321
VWR® Traceable® Humidity Sponge™, Dessicants
Desiccators Desiccants
Humidity Sponges™ meet FDA (Food and Drug Administration) requirements for use in direct contact with food and drugs. Desiccant (sodium calcium aluminosilicate hydrate) is non toxic and is a GRAS (Generally Recognised as Safe) material. It is ideal for storing food/pharmaceutical samples, electronics, PC boards, optics, chemicals and equipment. Outer material is made from DuPont’s Tyvek® breathable, high-density polyethylene.
  • Drying capability depends on volume, temperature, and moisture
  • Bags are guaranteed to retain 16% moisture by weight, typical results
More Product Information