VWR® Traceable®, Digital refrigerator and freezer thermometers
Supplier: VWR Collection
61161-364
VWRU61161-364EA 179 SGD
VWRU61161-364
VWR® Traceable®, Digital refrigerator and freezer thermometers
Thermometers Digital Thermometers
These thermometers accurately monitor temperatures in freezers, water baths, heating blocks, incubators, and refrigerators with this enclosed temperature-buffered sensor.
  • Triple display simultaneously shows min/max and current temperatures, °F/°C switchable
  • Bottle insulates sensor from rapid temperature changes when refrigerator door is opened
  • Ten-foot micro-cable permits refrigerator doors to close on it
  • High and low settable alarms
Non toxic glycol bottle probe.
Certifications: Traceable to NIST. Calibration compliant with ISO/IEC 17025, ANSI/More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable